7-Bromo-4-fluoro-2,3-dihydro-1H-inden-1-one 95%

Supplier: Ambeed
Warning

A430103-100MG A430103-10G A430103-1G A430103-250MG A430103-5G
76829-520EA 9 USD
76829-520 76829-522 76829-524 76829-526 76829-528
7-Bromo-4-fluoro-2,3-dihydro-1H-inden-1-one 95%
7-Bromo-4-fluoro-2,3-dihydro-1H-inden-1-one

Precaución: for Research Use Only

Formula: C₉H₆BrFO
MDL Number: MFCD17677341
CAS Number: 881189-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR