Methyl 5-bromo-6-fluoro-1H-indazole-4-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A428187-100MG
A428187-1G
A428187-250MG
76829-512EA
18
USD
76829-512
76829-514
76829-516
Methyl 5-bromo-6-fluoro-1H-indazole-4-carboxylate 95%
Methyl 5-bromo-6-fluoro-1H-indazole-4-carboxylate
Precaución: for Research Use Only
Formula:
C₉H₆BrFN₂O₂ |
MDL Number:
MFCD13177016 CAS Number: 1037841-25-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...