7-Amino-2H-1,4-benzoxazin-3(4H)-one 96%

Supplier: Ambeed
Warning

Synonyms: 7-Amino-2H-1,4-benzoxazin-3[4H]one

A427536-100MG A427536-1G A427536-5G A427536-250MG
76829-498EA 23 USD
76829-498 76829-500 76829-502 77886-134
7-Amino-2H-1,4-benzoxazin-3(4H)-one 96%
7-Amino-2H-1,4-benzoxazin-3(4H)-one

Precaución: for Research Use Only

Formula: C₈H₈N₂O₂
MW: 164.17 g/mol
MDL Number: MFCD03425794
CAS Number: 26215-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR