2,2-Difluoro-5-(hydroxymethyl)-1,3-benzodioxole 98%

Supplier: Ambeed
Warning

Synonyms: (2,2-Difluorobenzo[d][1,3]dioxol-5-yl)methanol, (2,2-Difluoro-1,3-benzodioxol-5-yl)methanol

A427027-1G A427027-25G
76829-494EA 49.87 USD
76829-494 76829-496
2,2-Difluoro-5-(hydroxymethyl)-1,3-benzodioxole 98%
2,2-Difluoro-5-(hydroxymethyl)-1,3-benzodioxole

Precaución: for Research Use Only

Formula: C₈H₆F₂O₃
MW: 188.128 g/mol
MDL Number: MFCD06202699
CAS Number: 72768-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR