4-(Hydroxymethyl)-2-isopropylthiazole 98%

Supplier: Ambeed
Warning

Synonyms: (2-Isopropylthiazol-4-yl)methanol

A425664-100MG A425664-1G A425664-250MG A425664-5G
76829-484EA 31.83 USD
76829-484 76829-486 76829-488 76829-490
4-(Hydroxymethyl)-2-isopropylthiazole 98%
4-(Hydroxymethyl)-2-isopropylthiazole

Precaución: for Research Use Only

Formula: C₇H₁₁NOS
MW: 157.24 g/mol
MDL Number: MFCD09870064
CAS Number: 133047-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR