Creatine phosphate disodium salt hydrate 98%

Supplier: Ambeed
Warning

Synonyms: Phosphocreatine disodium hydrate, Sodium 2-(1-methyl-3-phosphonatoguanidino)acetate hydrate(2:1:x), N-(Imino[phosphonoamino]methyl)-N-methylglycine hydrate, Phosphocreatine disodium salt hydrate

A424009-1G A424009-25G A424009-5G A424009-100G
76829-440EA 5.31 USD
76829-440 76829-442 77613-312 77684-182
Creatine phosphate disodium salt hydrate 98%
Creatine phosphate disodium salt hydrate

Precaución: For research use only.

Formula: C₄H₈N₃Na₂O₅P·H₂O
MW: 273.09 g/mol
MDL Number: MFCD00150191
CAS Number: 19333-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR