Methyl 6-chloro-5-(trifluoromethyl)picolinate 97%

Supplier: Ambeed
Warning

A419238-100MG A419238-1G A419238-250MG
76829-432EA 16 USD
76829-432 76829-434 76829-436
Methyl 6-chloro-5-(trifluoromethyl)picolinate 97%
Methyl 6-chloro-5-(trifluoromethyl)picolinate

Precaución: for Research Use Only

Formula: C₈H₅ClF₃NO₂
MW: 239.57 g/mol
MDL Number: MFCD18255501
CAS Number: 1211518-35-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR