Methyl 3-(benzyloxy)-5-hydroxybenzoate 97%

Supplier: Ambeed
Warning

A418333-10G A418333-1G A418333-250MG A418333-5G
76829-418EA 137 USD
76829-418 76829-420 76829-422 76829-424
Methyl 3-(benzyloxy)-5-hydroxybenzoate 97%
Methyl 3-(benzyloxy)-5-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₄
MDL Number: MFCD06655985
CAS Number: 54915-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR