N1,N1,N6,N6-Tetramethyladipamide 95%

Supplier: Ambeed
Warning

A418095-100MG A418095-1G A418095-250MG A418095-5G
76829-410EA 9 USD
76829-410 76829-412 76829-414 76829-416
N1,N1,N6,N6-Tetramethyladipamide 95%
N1,N1,N6,N6-Tetramethyladipamide

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
MDL Number: MFCD01753239
CAS Number: 3644-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR