(S)-3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid 98%

Supplier: Ambeed
Warning

Synonyms: (S)-3-((tert-Butoxycarbonyl)amino)-3-(3-methoxyphenyl)propanoic acid

A418030-100MG A418030-1G A418030-250MG A418030-5G
76829-404EA 33.95 USD
76829-404 76829-406 76829-408 77684-170
(S)-3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid 98%
(S)-3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid

Precaución: For research use only.

Formula: C₁₅H₂₁NO₅
MW: 295.3309 g/mol
MDL Number: MFCD03427899
CAS Number: 499995-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR