2-Methoxy-3-methylaniline 97%

Supplier: Ambeed
Warning

A416442-1G A416442-250MG A416442-25G A416442-5G
76829-394EA 53.05 USD
76829-394 76829-396 76829-398 76829-400
2-Methoxy-3-methylaniline 97%
2-Methoxy-3-methylaniline

Precaución: for Research Use Only

Formula: C₈H₁₁NO
MW: 137.179 g/mol
MDL Number: MFCD12547890
CAS Number: 18102-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR