3-(trans-4-Hydroxycyclohexylcarbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (3-((trans-4-Hydroxycyclohexyl)carbamoyl)phenyl)boronic acid

A415505-100MG A415505-1G A415505-250MG
76829-388EA 37.14 USD
76829-388 76829-390 76829-392
3-(trans-4-Hydroxycyclohexylcarbamoyl)phenylboronic acid 98%
3-(trans-4-Hydroxycyclohexylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₈BNO₄
MW: 263.0973 g/mol
MDL Number: MFCD09878335
CAS Number: 957062-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR