6-Benzyladenine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 6-Benzylaminopurine hydrochloride, N-Benzyl-9H-purin-6-amine hydrochloride

A414846-1G A414846-5G A414846-250MG
76829-372EA 29.71 USD
76829-372 76829-374 77572-290
6-Benzyladenine hydrochloride 98%
6-Benzyladenine hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₂ClN₅
MW: 261.71 g/mol
MDL Number: MFCD09265275
CAS Number: 162714-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR