2-Bromo-1-(1-methyl-1H-pyrazol-4-yl)ethanone 95%

Supplier: Ambeed
Danger

A413545-250MG A413545-1G
76829-364EA 182.48 USD
76829-364 77613-264
2-Bromo-1-(1-methyl-1H-pyrazol-4-yl)ethanone 95%
2-Bromo-1-(1-methyl-1H-pyrazol-4-yl)ethanone

Precaución: for Research Use Only

Formula: C₆H₇BrN₂O
MW: 203.03 g/mol
MDL Number: MFCD11182388
CAS Number: 706819-66-1
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR