4,4'-Di(4-pyridyl)biphenyl 98%

Supplier: Ambeed
Warning

Synonyms: 4,4'-Bis(4-pyridyl)biphenyl, 4,4'-Di(pyridin-4-yl)-1,1'-biphenyl

A413005-5G A413005-1G A413005-250MG
76829-362EA 594.11 USD
76829-362 77684-158 77684-159
4,4'-Di(4-pyridyl)biphenyl 98%
4,4'-Di(4-pyridyl)biphenyl

Precaución: For research use only.

Formula: C₂₂H₁₆N₂
MW: 308.38 g/mol
MDL Number: MFCD11977635
CAS Number: 319430-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR