3-Chloro-4-(1H-imidazol-1-yl)benzaldehyde 97%

Supplier: Ambeed
Warning

A411437-1G A411437-5G
76829-352EA 24 USD
76829-352 76829-354
3-Chloro-4-(1H-imidazol-1-yl)benzaldehyde 97%
3-Chloro-4-(1H-imidazol-1-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₇ClN₂O
MW: 206.63 g/mol
MDL Number: MFCD11656796
CAS Number: 870837-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR