3-Bromo-6-fluoropyridine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-2-carboxy-6-fluoropyridine, 3-Bromo-6-fluoropicolinic acid

A409699-100MG A409699-1G A409699-250MG A409699-5G
76829-338EA 48.81 USD
76829-338 76829-340 76829-342 77551-226
3-Bromo-6-fluoropyridine-2-carboxylic acid 95%
3-Bromo-6-fluoropyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₃BrFNO₂
MW: 219.996 g/mol
MDL Number: MFCD13185802
CAS Number: 1211589-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR