1-(3-Fluorophenyl)cyclopropanecarbonitrile 95%

Supplier: Ambeed
Warning

A409306-100MG A409306-1G A409306-250MG
76829-328EA 46.68 USD
76829-328 76829-330 76829-332
1-(3-Fluorophenyl)cyclopropanecarbonitrile 95%
1-(3-Fluorophenyl)cyclopropanecarbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₈FN
MW: 161.17 g/mol
MDL Number: MFCD07374415
CAS Number: 124276-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR