2-(tert-Butyldimethylsilyloxy)-4-bromoanisole 98%

Supplier: Ambeed
Warning

Synonyms: (5-Bromo-2-methoxyphenoxy)(tert-butyl)dimethylsilane

A407694-1G A407694-5G
76829-316EA 39.26 USD
76829-316 76829-318
2-(tert-Butyldimethylsilyloxy)-4-bromoanisole 98%
2-(tert-Butyldimethylsilyloxy)-4-bromoanisole

Precaución: for Research Use Only

Formula: C₁₃H₂₁BrO₂Si
MW: 317.2941 g/mol
MDL Number: MFCD11855959
CAS Number: 177329-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR