4-Bromo-3-chloro-2-fluoroiodobenzene 97%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-2-chloro-3-fluoro-4-iodobenzene

A407484-1G A407484-25G A407484-5G
76829-310EA 35.01 USD
76829-310 76829-312 76829-314
4-Bromo-3-chloro-2-fluoroiodobenzene 97%
4-Bromo-3-chloro-2-fluoroiodobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrClFI
MW: 335.34 g/mol
MDL Number: MFCD09878167
CAS Number: 1000573-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR