2-Amino-3-hydroxy-3-(thiophen-2-yl)propanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: H-β-(2-Thienyl)-DL-Ser-OH, H-DL-β-(2-Thienyl)-DL-Ser-OH

A406989-250MG A406989-1G A406989-5G
76829-306EA 20.16 USD
76829-306 77572-260 77684-148
2-Amino-3-hydroxy-3-(thiophen-2-yl)propanoic acid 98%
2-Amino-3-hydroxy-3-(thiophen-2-yl)propanoic acid

Precaución: For research use only.

Formula: C₇H₉NO₃S
MW: 187.213 g/mol
MDL Number: MFCD00067304
CAS Number: 32595-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR