4-Methyl-2-nitrobenzylalcohol 97%

Supplier: Ambeed
Warning

Synonyms: (4-Methyl-2-nitrophenyl)methanol

A406413-100MG A406413-1G A406413-250MG
76829-294EA 27.59 USD
76829-294 76829-296 76829-298
4-Methyl-2-nitrobenzylalcohol 97%
4-Methyl-2-nitrobenzylalcohol

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.162 g/mol
MDL Number: MFCD08275452
CAS Number: 22996-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR