4-Bromo-1-methoxy-2-(3-methoxypropoxy)benzene 98%

Supplier: Ambeed
Warning

A404925-100G A404925-1G A404925-25G A404925-5G
76829-286EA 206 USD
76829-286 76829-288 76829-290 76829-292
4-Bromo-1-methoxy-2-(3-methoxypropoxy)benzene 98%
4-Bromo-1-methoxy-2-(3-methoxypropoxy)benzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrO₃
MDL Number: MFCD14582816
CAS Number: 173336-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR