4-Phenyl-4H-1,2,4-triazole 98%

Supplier: Ambeed
Warning

A403811-100MG A403811-1G A403811-250MG
76829-276EA 36 USD
76829-276 76829-278 76829-280
4-Phenyl-4H-1,2,4-triazole 98%
4-Phenyl-4H-1,2,4-triazole

Precaución: for Research Use Only

Formula: C₈H₇N₃
MW: 145.16 g/mol
MDL Number: MFCD00957679
CAS Number: 16227-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR