(R)-1-(4-Bromophenyl)-2,2,2-trifluoroethanamine 97%

Supplier: Ambeed
Warning

A400481-100MG A400481-1G A400481-250MG
76829-256EA 25 USD
76829-256 76829-258 77886-072
(R)-1-(4-Bromophenyl)-2,2,2-trifluoroethanamine 97%
(R)-1-(4-Bromophenyl)-2,2,2-trifluoroethanamine

Precaución: for Research Use Only

Formula: C₈H₇BrF₃N
MDL Number: MFCD07374594
CAS Number: 843608-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR