4-Benzoylphenyl acrylate 98%

Supplier: Ambeed
Warning

A398305-100G A398305-10G A398305-1G A398305-250MG A398305-25G A398305-5G
76829-236EA 54 USD
76829-236 76829-238 76829-240 76829-242 76829-244 77886-065
4-Benzoylphenyl acrylate 98%
4-Benzoylphenyl acrylate

Precaución: for Research Use Only

Formula: C₁₆H₁₂O₃
MDL Number: MFCD12828285
CAS Number: 22535-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR