2-Anilino-9,9-dimethylfluorene 97%

Supplier: Ambeed
Warning

Synonyms: 9,9-Dimethyl-N-phenyl-9H-fluoren-2-amine

A398266-1G A398266-5G A398266-25G
76829-230EA 9 USD
76829-230 76829-232 76829-234 77886-064
2-Anilino-9,9-dimethylfluorene 97%
2-Anilino-9,9-dimethylfluorene

Precaución: for Research Use Only

Formula: C₂₁H₁₉N
MW: 285.39 g/mol
MDL Number: MFCD20040462
CAS Number: 355832-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR