Ethyl 2,2-difluoro-2-(4-nitrophenyl)acetate 95%

Supplier: Ambeed
Warning

A398258-1G A398258-250MG A398258-5G
76829-224EA 127.31 USD
76829-224 76829-226 76829-228
Ethyl 2,2-difluoro-2-(4-nitrophenyl)acetate 95%
Ethyl 2,2-difluoro-2-(4-nitrophenyl)acetate

Precaución: for Research Use Only

Formula: C₁₀H₉F₂NO₄
MW: 245.17 g/mol
MDL Number: MFCD11975616
CAS Number: 243656-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR