1-Iodo-2,4-dimethyl-3-nitrobenzene 97%

Supplier: Ambeed
Warning

A397207-100MG A397207-1G A397207-250MG A397207-5G A397207-25G
76829-210EA 11 USD
76829-210 76829-212 76829-214 76829-216 77886-059
1-Iodo-2,4-dimethyl-3-nitrobenzene 97%
1-Iodo-2,4-dimethyl-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₈H₈INO₂
MDL Number: MFCD02709656
CAS Number: 56404-21-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR