1,4,7-Tris(ethoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane 98%

Supplier: Ambeed
Warning

Synonyms: Triethyl 2,2',2''-(1,4,7,10-tetraazacyclododecane-1,4,7-triyl)triacetate

A397202-1G A397202-250MG
76829-206EA 74.27 USD
76829-206 76829-208
1,4,7-Tris(ethoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane 98%
1,4,7-Tris(ethoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane

Precaución: for Research Use Only

Formula: C₂₀H₃₈N₄O₆
MW: 430.55 g/mol
MDL Number: MFCD09263320
CAS Number: 114873-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR