Methyl 4,5-dichlorothiophene-2-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A394249-100MG
A394249-10G
A394249-1G
A394249-250MG
A394249-5G
76829-156EA
35.01
USD
76829-156
76829-158
76829-160
76829-162
76829-164
Methyl 4,5-dichlorothiophene-2-carboxylate 95%
Methyl 4,5-dichlorothiophene-2-carboxylate
Precaución: for Research Use Only
Formula:
C₆H₄Cl₂O₂S MW: 211.06 g/mol |
MDL Number:
MFCD16038671 CAS Number: 89281-29-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...