(R)-3-Aminopyrrolidin-2-one hydrochloride 95%

Supplier: Ambeed
Warning

A391752-1G A391752-100MG A391752-250MG A391752-5G
76829-148EA 182 USD
76829-148 77886-038 77886-039 77886-040
(R)-3-Aminopyrrolidin-2-one hydrochloride 95%
(R)-3-Aminopyrrolidin-2-one hydrochloride

Precaución: for Research Use Only

Formula: C₄H₈N₂O·ClH
MDL Number: MFCD14635509
CAS Number: 223407-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR