2-Chlorobenzo[d]thiazole-5-carbonitrile 97%

Supplier: Ambeed
Warning

A390819-1G A390819-250MG
76829-134EA 56 USD
76829-134 76829-136
2-Chlorobenzo[d]thiazole-5-carbonitrile 97%
2-Chlorobenzo[d]thiazole-5-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₃ClN₂S
MDL Number: MFCD12827959
CAS Number: 385432-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR