3-(1,2,4-Triazol-3-yl)carbamoyl)phenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (3-((1H-1,2,4-Triazol-3-yl)carbamoyl)phenyl)boronic acid

A390170-1G A390170-250MG A390170-5G
76829-128EA 166 USD
76829-128 76829-130 76829-132
3-(1,2,4-Triazol-3-yl)carbamoyl)phenylboronic acid 95%
3-(1,2,4-Triazol-3-yl)carbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₉BN₄O₃
MDL Number: MFCD07363798
CAS Number: 871333-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR