4-Chloro-2-(methylthio)-5,6,7,8-tetrahydroquinazoline 97%

Supplier: Ambeed
Warning

A390127-100MG A390127-1G A390127-250MG
76829-122EA 15.92 USD
76829-122 76829-124 76829-126
4-Chloro-2-(methylthio)-5,6,7,8-tetrahydroquinazoline 97%
4-Chloro-2-(methylthio)-5,6,7,8-tetrahydroquinazoline

Precaución: for Research Use Only

Formula: C₉H₁₁ClN₂S
MW: 214.72 g/mol
MDL Number: MFCD01568403
CAS Number: 51660-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR