(1-Tetradecyl)triphenylphosphonium bromide 97%

Supplier: Ambeed
Warning

Synonyms: Triphenyl(tetradecyl)phosphonium bromide, Tetradecyltriphenylphosphonium bromide

A388943-1G A388943-25G A388943-5G
76829-096EA 5 USD
76829-096 76829-098 76829-100
(1-Tetradecyl)triphenylphosphonium bromide 97%
(1-Tetradecyl)triphenylphosphonium bromide

Precaución: for Research Use Only

Formula: H₃C(CH₂)₁₃P(C₆H₅)₃Br
MDL Number: MFCD00051857
CAS Number: 25791-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR