3-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester 98%

Supplier: Ambeed
Danger

Synonyms: 2-(3-(Bromomethyl)-2-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A388020-1G A388020-250MG A388020-5G
76829-086EA 72.15 USD
76829-086 76829-088 76829-090
3-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester 98%
3-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BBrFO₂
MW: 314.989 g/mol
MDL Number: MFCD17214291
CAS Number: 1256360-37-8
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR