2-Bromo-5,8-dioxaspiro[3.4]octane 98%

Supplier: Ambeed
Warning

A387723-1G A387723-250MG A387723-100MG
76829-084EA 66 USD
76829-084 77886-024 77886-023
2-Bromo-5,8-dioxaspiro[3.4]octane 98%
2-Bromo-5,8-dioxaspiro[3.4]octane

Precaución: for Research Use Only

Formula: C₆H₉BrO₂
MDL Number: MFCD20637976
CAS Number: 1257996-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR