2-[2-(Vinyloxy)ethoxy]isoindoline-1,3-dione 95% stabilized

Supplier: Ambeed
Warning

Synonyms: 2-(2-(Vinyloxy)ethoxy)isoindoline-1,3-dione

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A384646-100MG A384646-1G A384646-250MG
76829-056EA 37.14 USD
76829-056 76829-058 76829-060
2-[2-(Vinyloxy)ethoxy]isoindoline-1,3-dione 95% stabilized
2-[2-(Vinyloxy)ethoxy]isoindoline-1,3-dione

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO₄
MW: 233.22 g/mol
MDL Number: MFCD15142583
CAS Number: 391212-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR