4-(2-(Piperidin-1-yl)ethoxy)phenylboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 1-(2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)ethyl)piperidine

A384011-100MG A384011-1G A384011-250MG
76829-050EA 14 USD
76829-050 76829-052 76829-054
4-(2-(Piperidin-1-yl)ethoxy)phenylboronic acid pinacol ester 95%
4-(2-(Piperidin-1-yl)ethoxy)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₉H₃₀BNO₃
MDL Number: MFCD09258652
CAS Number: 934586-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR