2-Chloro-5-methoxy-4-pyridineboronic acid 98%

Supplier: Ambeed
Danger

Synonyms: (2-Chloro-5-methoxypyridin-4-yl)boronic acid, 2-Chloro-5-methoxypyridine-4-boronic acid

A383542-100MG A383542-1G A383542-250MG
76829-044EA 30.77 USD
76829-044 76829-046 76829-048
2-Chloro-5-methoxy-4-pyridineboronic acid 98%
2-Chloro-5-methoxy-4-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₇BClNO₃
MW: 187.39 g/mol
MDL Number: MFCD09972106
CAS Number: 1072945-98-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR