2-(Bromomethyl)-4-fluorophenylboronic acid pinacol ester 98%

Supplier: Ambeed
Danger

Synonyms: 2-(2-(Bromomethyl)-4-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A383410-1G A383410-250MG A383410-5G
76829-032EA 94.43 USD
76829-032 76829-034 76829-036
2-(Bromomethyl)-4-fluorophenylboronic acid pinacol ester 98%
2-(Bromomethyl)-4-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BBrFO₂
MW: 314.9863 g/mol
MDL Number: MFCD06659927
CAS Number: 850567-57-6
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR