Methyl 4-hydroxypiperidine-4-carboxylate hydrochloride 97%

Supplier: Ambeed
Warning

A382657-1G A382657-250MG A382657-5G
76829-026EA 1159.57 USD
76829-026 76829-028 76829-030
Methyl 4-hydroxypiperidine-4-carboxylate hydrochloride 97%
Methyl 4-hydroxypiperidine-4-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₄ClNO₃
MW: 195.64 g/mol
MDL Number: MFCD11845425
CAS Number: 179022-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR