Ethylhydrazine hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A381958-10G
A381958-1G
A381958-25G
A381958-5G
A381958-250MG
76829-018EA
412.7
USD
76829-018
76829-020
76829-022
76829-024
77684-100
Ethylhydrazine hydrochloride 95%
Ethylhydrazine hydrochloride
Precaución: For research use only.
Formula:
C₂H₈N₂·ClH MW: 96.559 g/mol |
MDL Number:
MFCD01722460 CAS Number: 18413-14-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...