3-Chloro-5-iodopyridin-2-amine 98%

Supplier: Ambeed
Warning

A381314-1G A381314-250MG A381314-5G
76829-008EA 92.3 USD
76829-008 76829-010 76829-012
3-Chloro-5-iodopyridin-2-amine 98%
3-Chloro-5-iodopyridin-2-amine

Precaución: for Research Use Only

Formula: C₅H₄ClIN₂
MW: 254.45 g/mol
MDL Number: MFCD09754395
CAS Number: 952901-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR