Methyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-Chloro-5-(trifluoromethyl)pyridine-2-carboxylate, 3-Chloro-2-(methoxycarbonyl)-5-(trifluoromethyl)pyridine, Methyl 3-chloro-5-(trifluoromethyl)picolinate

A380894-1G A380894-25G A380894-5G
76829-002EA 14 USD
76829-002 76829-004 76829-006
Methyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate 98%
Methyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₈H₅ClF₃NO₂
MW: 239.579 g/mol
MDL Number: MFCD01570523
CAS Number: 655235-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR