(2,6-Dibromopyridin-3-yl)methanol 98%

Supplier: Ambeed
Warning

A379075-100MG A379075-1G A379075-250MG
76828-986EA 11 USD
76828-986 76828-988 76828-990
(2,6-Dibromopyridin-3-yl)methanol 98%
(2,6-Dibromopyridin-3-yl)methanol

Precaución: for Research Use Only

Formula: C₆H₅Br₂NO
MDL Number: MFCD02942305
CAS Number: 55483-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR