Bis(cycloocta-1,5-diene)rhodium(I) hexafluorophosphate 98%

Supplier: Ambeed
Warning

A378330-100MG A378330-1G A378330-250MG A378330-50MG
76828-974EA 171 USD
76828-974 76828-976 76828-978 76828-980
Bis(cycloocta-1,5-diene)rhodium(I) hexafluorophosphate 98%
Bis(cycloocta-1,5-diene)rhodium(I) hexafluorophosphate

Precaución: for Research Use Only

Formula: C₁₆H₂₄F₆PRh
MDL Number: MFCD26793487
CAS Number: 62793-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR