3-Methoxy-2-naphthaldehyde 98%

Supplier: Ambeed
Warning

A377005-250MG A377005-5G A377005-1G A377005-25G
76828-948EA 7 USD
76828-948 76828-950 77885-997 77885-998
3-Methoxy-2-naphthaldehyde 98%
3-Methoxy-2-naphthaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₂
MDL Number: MFCD13191799
CAS Number: 56679-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR