(4-(Naphthalen-1-yl(phenyl)amino)phenyl)boronic acid 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A376168-100MG A376168-250MG A376168-1G
76828-938EA 24 USD
76828-938 76828-940 77684-083
(4-(Naphthalen-1-yl(phenyl)amino)phenyl)boronic acid 95%, Ambeed.Inc
(4-(Naphthalen-1-yl(phenyl)amino)phenyl)boronic acid

Precaución: For research use only.

Formula: C₂₂H₁₈BNO₂
MW: 339.20 g/mol
MDL Number: MFCD22493467
CAS Number: 717888-41-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR